siRNA Details
Gene Symbol: Zfp458 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001152.2 | NP_001001152.1 | 4 | 3000 | ||
GenBank mRNA | AK133308.1 | - | 2919 | |||
AK161458.1 | - | 2154 | ||||
BC062958.1 | - | 4 | 3000 | 30548236 |
