siRNA Details
Gene Symbol: BC031181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001181.3 | NP_001001181.1 | 3 | 552 | ||
GenBank mRNA | AK031999.1 | - | 2 | 226 | ||
AK077642.1 | - | 3 | 485 | |||
AK131658.1 | - | 3 | 549 | |||
AK216918.1 | - | 2 | 195 | |||
BC016084.1 | - | 3 | 552 | 4501468 | ||
BC027819.1 | - | 3 | 566 | 5320851 | ||
BC031181.1 | - | 3 | 547 | 4988367 |
