siRNA Details
Gene Symbol: Iqgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033484.1 | NP_001028656.1 | 4 | 321 | ||
GenBank mRNA | AK050913.1 | - | 3 | 1325 | ||
AK162700.1 | - | 4 | 344 | |||
BC068159.1 | - | 4 | 321 | |||
BC151102.1 | - | 4 | 401 | 9088015 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
