siRNA Details
Gene Symbol: Cyp2c44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001446.3 | NP_001001446.3 | 5 | 731 | ||
NM_001167905.1 | NP_001161377.1 | 5 | 731 | |||
GenBank mRNA | AK050436.1 | - | 5 | 693 | ||
AK149451.1 | - | 5 | 693 | |||
BC022141.1 | - | 5 | 680 | |||
BC024068.1 | - | 5 | 651 | |||
BC025819.1 | - | 5 | 683 | 5099385 | ||
BC026766.1 | - | 5 | 683 | 4195655 | ||
BC034834.1 | - | 5 | 676 | 5099935 |
