siRNA Details
Gene Symbol: Tpm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001491.1 | NP_001001491.1 | 8 | 792 | ||
GenBank mRNA | AK206089.1 | - | 5 | 328 | ||
AK207394.1 | - | 5 | 325 | |||
BC023701.1 | - | 8 | 799 | 5345587 | ||
BC023827.1 | - | 8 | 786 | 5343662 | ||
BC032174.1 | - | 8 | 788 | |||
BC032175.1 | - | 8 | 788 | |||
BC070421.1 | - | 8 | 792 | 6403300 |
