siRNA Details
Gene Symbol: Tpm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001491.1 | NP_001001491.1 | 8 | 1048 | ||
GenBank mRNA | BC023701.1 | - | 8 | 1055 | 5345587 | |
BC023827.1 | - | 8 | 1042 | 5343662 | ||
BC032174.1 | - | 8 | 1044 | |||
BC032175.1 | - | 8 | 1044 | |||
BC070421.1 | - | 8 | 1048 | 6403300 |
