siRNA Details
Gene Symbol: LINC00898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033377.1 | 2, 3 | 2390 | |||
GenBank mRNA | AK128136.1 | - | 2, 3 | 2387 | ||
BC132762.1 | - | 2, 3 | 723 | |||
BC132788.1 | - | 2, 3 | 723 |
