siRNA Details
Gene Symbol: FLJ33360 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028351.1 | 2 | 1350 | |||
GenBank mRNA | AK090679.1 | - | 2 | 1350 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FLJ33360 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028351.1 | 2 | 1350 | |||
GenBank mRNA | AK090679.1 | - | 2 | 1350 |