siRNA Details
Gene Symbol: CNKSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168648.1 | NP_001162119.1 | 21 | 3798 | ||
NM_001168649.1 | NP_001162120.1 | 20 | 3651 | |||
GenBank mRNA | AB020709.1 | - | 21 | 3773 | ||
AK056549.1 | - | 10 | 2150 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
