siRNA Details
Gene Symbol: MIR4697HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024344.1 | 1 | 3267 | |||
GenBank mRNA | AK123849.1 | - | 1 | 2271 | ||
AL133591.1 | - | 1 | 1196 | |||
BC106032.1 | - | 1 | 3128 | |||
BX641055.1 | - | 1 | 2888 |
