siRNA Details
Gene Symbol: OTUD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102653.1 | NP_001096123.1 | 21 | 6544 | ||
XM_005263079.4 | XP_005263136.1 | 21 | 6647 | |||
XM_011532040.2 | XP_011530342.2 | 21 | 6648 | |||
XM_011532041.2 | XP_011530343.2 | 21 | 6645 | |||
XM_011532042.2 | XP_011530344.1 | 21 | 6569 | |||
GenBank mRNA | AB028969.1 | - | 1 | 5054 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
