siRNA Details
Gene Symbol: Hecw2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001883.3 | NP_001001883.1 | 8 | 987 | ||
GenBank mRNA | AB182244.1 | - | 7 | 893 | ||
AK129325.1 | - | 6 | 524 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
