siRNA Details
Gene Symbol: Hecw2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001883.3 | NP_001001883.1 | 9, 10 | 2458 | ||
GenBank mRNA | AB182244.1 | - | 8, 9 | 2364 | ||
AK129325.1 | - | 7, 8 | 1995 |

More Information
Other Species Targeted

Gene Ontology Categories:
