siRNA Details
Gene Symbol: 3110053B16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014209.1 | - | 2 | 767 | ||
AK148766.1 | - | 4 | 1103 |
Gene Symbol: 2410018L13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK010549.1 | - | 3 | 982 | ||
BC063063.1 | - | 4 | 1183 |
Gene Symbol: 6030426L16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020054.1 | - | 1160 |
Gene Symbol: Gm19924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC099486.1 | - | 2 | 636 | 30937101 |
Gene Symbol: Gm4983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AV212851.1 | - | 1 | 187 | ||
BC030401.1 | - | 1026 | 5362444 | |||
BC083083.1 | - | 884 | 6514154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
