siRNA Details
Gene Symbol: Atp6v1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK085469.1 | - | 2 | 881 | ||
BC013168.1 | - | 2 | 867 | |||
BC058584.1 | - | 2 | 872 | |||
BC080796.1 | - | 2 | 901 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
