siRNA Details
Gene Symbol: AU019823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134902.1 | NP_001128374.1 | 7 | 1683 | ||
NM_212449.2 | NP_997614.2 | 6 | 1656 | |||
GenBank mRNA | BC051390.1 | - | 1 | 703 | ||
BC067205.1 | - | 6 | 1438 | 6335744 |
