siRNA Details
Gene Symbol: Atp6v1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212490.2 | NP_997655.1 | 3 | 1285 | ||
XM_008772735.1 | XP_008770957.1 | 3 | 1076 | |||
XM_008772736.1 | XP_008770958.1 | 3 | 1056 | |||
GenBank mRNA | FQ212959.1 | - | 3 | 1247 | ||
FQ213756.1 | - | 3 | 1138 |
