siRNA Details
Gene Symbol: Vwa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212499.2 | NP_997664.1 | 15 | 2476 | ||
XM_008772728.1 | XP_008770950.1 | 14 | 2910 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vwa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212499.2 | NP_997664.1 | 15 | 2476 | ||
XM_008772728.1 | XP_008770950.1 | 14 | 2910 |