siRNA Details
Gene Symbol: Atp5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212516.2 | NP_997681.1 | 3 | 252 | ||
GenBank mRNA | BC058497.1 | - | 3 | 253 | 6918545 | |
FQ210248.1 | - | 3 | 242 | |||
FQ211867.1 | - | 3 | 249 | |||
FQ214576.1 | - | 248 | ||||
FQ217032.1 | - | 3 | 235 | |||
FQ217139.1 | - | 3 | 268 | |||
FQ217410.1 | - | 3 | 269 | |||
FQ217906.1 | - | 3 | 241 | |||
FQ218008.1 | - | 3 | 241 | |||
FQ221909.1 | - | 3 | 281 | |||
FQ223878.1 | - | 3 | 242 | |||
FQ223920.1 | - | 3 | 248 | |||
FQ224004.1 | - | 3 | 242 | |||
FQ224067.1 | - | 3 | 242 | |||
FQ224620.1 | - | 3 | 242 | |||
FQ224644.1 | - | 3 | 248 | |||
FQ229120.1 | - | 3 | 284 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013795, BC094407, AK009465, AK008012, Y17223, and others.
Gene Ontology Categories:
