siRNA Details
Gene Symbol: Abhd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212524.1 | NP_997689.1 | 3, 4 | 560 | ||
XM_006244107.1 | XP_006244169.1 | 4, 5 | 665 | |||
XM_006244108.2 | XP_006244170.1 | 3, 4 | 565 | |||
XM_006244109.2 | XP_006244171.1 | 2, 3 | 1953 | |||
GenBank mRNA | AY550934.1 | - | 3, 4 | 560 | ||
BC127470.1 | - | 3, 4 | 594 |
