siRNA Details
Gene Symbol: Abhd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212524.1 | NP_997689.1 | 7 | 1030 | ||
XM_006244107.1 | XP_006244169.1 | 8 | 1135 | |||
XM_006244108.2 | XP_006244170.1 | 7 | 1035 | |||
XM_006244109.2 | XP_006244171.1 | 6 | 2423 | |||
GenBank mRNA | AY550934.1 | - | 7 | 1030 | ||
BC127470.1 | - | 7 | 1064 |
