siRNA Details
Gene Symbol: Ddah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165936.1 | NP_001159408.1 | 6 | 857 | ||
NM_212532.2 | NP_997697.1 | 6 | 904 | |||
XM_008772714.1 | XP_008770936.1 | 7 | 1003 | |||
XM_008772715.1 | XP_008770937.1 | 6 | 1117 | |||
GenBank mRNA | BC086443.1 | - | 6 | 805 | 7303579 | |
BC097930.1 | - | 6 | 894 | 7388716 |
