siRNA Details
Gene Symbol: Phf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212538.1 | NP_997703.1 | 3 | 424 | ||
XM_006256117.2 | XP_006256179.1 | 3 | 678 | |||
XM_008772723.1 | XP_008770945.1 | 3 | 679 | |||
GenBank mRNA | BC083843.1 | - | 3 | 365 | 7114670 |
