siRNA Details
Gene Symbol: Rgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212547.3 | NP_997712.1 | 8, 9 | 1251 | ||
XM_008772722.1 | XP_008770944.1 | 8, 9 | 1835 | |||
GenBank mRNA | BC107439.1 | - | 8, 9 | 1222 | 7121126 | |
FQ211603.1 | - | 8, 9 | 1257 |
