siRNA Details
Gene Symbol: Rgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212547.3 | NP_997712.1 | 2, 3 | 283 | ||
XM_008772722.1 | XP_008770944.1 | 2, 3 | 867 | |||
GenBank mRNA | BC107439.1 | - | 2, 3 | 254 | 7121126 | |
DY316563.1 | - | 2, 3 | 283 | |||
FQ211603.1 | - | 2, 3 | 275 |
