siRNA Details
Gene Symbol: Ring1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212549.2 | NP_997714.2 | 7 | 1544 | ||
GenBank mRNA | BC098635.1 | - | 7 | 1488 | 7098264 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ring1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212549.2 | NP_997714.2 | 7 | 1544 | ||
GenBank mRNA | BC098635.1 | - | 7 | 1488 | 7098264 |