siRNA Details
Gene Symbol: BOLA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212552.2 | NP_997717.2 | 3, 4 | 289 | ||
GenBank mRNA | BC017744.2 | - | 2 | 125 | ||
BC042036.1 | - | 3, 4 | 285 | |||
BU600037.1 | - | 294 | ||||
DQ225187.1 | - | 3, 4 | 251 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
