siRNA Details
Gene Symbol: NMNAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015039.3 | NP_055854.1 | 2 | 335 | ||
NM_170706.3 | NP_733820.1 | 2 | 223 | |||
GenBank mRNA | AB007948.1 | - | 2 | 203 | ||
AF288395.1 | - | 2 | 440 | |||
AK291390.1 | - | 4 | 335 | |||
BC020998.2 | - | 2 | 223 | 2824280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
