siRNA Details
Gene Symbol: Phactr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214458.1 | NP_999623.1 | 4 | 407 | ||
XM_008758610.1 | XP_008756832.1 | 4 | 1327 | |||
XM_008758611.1 | XP_008756833.1 | 4 | 1325 | |||
GenBank mRNA | AY500158.1 | - | 4 | 407 |
