siRNA Details
Gene Symbol: Sirt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008368.1 | NP_001008369.1 | 13 | 1276 | ||
XM_006228672.2 | XP_006228734.1 | 12 | 1224 | |||
XM_006228673.2 | XP_006228735.1 | 13 | 1229 | |||
XM_006228675.1 | XP_006228737.1 | 13 | 1008 | |||
XM_006228676.2 | XP_006228738.1 | 14 | 1013 | |||
GenBank mRNA | BC086545.1 | - | 13 | 1276 | 7134377 | |
FQ212805.1 | - | 13 | 935 |
