siRNA Details
Gene Symbol: Slc25a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057102.1 | NP_476443.1 | 2 | 588 | ||
GenBank mRNA | BC059108.1 | - | 2 | 637 | 6922359 | |
D12771.1 | - | 2 | 588 | |||
FQ213290.1 | - | 2 | 610 | |||
FQ213647.1 | - | 2 | 610 | |||
FQ214507.1 | - | 2 | 565 | |||
FQ219677.1 | - | 2 | 610 | |||
FQ220249.1 | - | 2 | 612 | |||
FQ220272.1 | - | 2 | 612 | |||
FQ220435.1 | - | 2 | 632 | |||
FQ229325.1 | - | 2 | 613 | |||
FQ229438.1 | - | 2 | 613 |
