siRNA Details
Gene Symbol: Wdr26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145514.5 | NP_663489.4 | 14 | 2742 | ||
GenBank mRNA | AK082326.1 | - | 15 | 2037 | ||
AK139350.1 | - | 1 | 293 | |||
AK167017.1 | - | 3 | 551 | |||
AK167116.1 | - | 3 | 551 | |||
BC094233.1 | - | 11 | 1626 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001109081, FQ226610, FQ227016.
Gene Ontology Categories:
