siRNA Details
Gene Symbol: Olfr530 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146519.1 | NP_666730.1 | 1 | 23 | ||
GenBank mRNA | BC138081.1 | - | 1 | 81 | 8861100 | |
BC138082.1 | - | 1 | 81 | 8861101 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
