siRNA Details
Gene Symbol: Celf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244891.1 | NP_001231820.1 | 15 | 7386 | ||
NM_001244903.1 | NP_001231832.1 | 14 | 7333 | |||
NM_017368.3 | NP_059064.2 | 15 | 4410 | |||
NM_198683.2 | NP_941955.1 | 14 | 4258 | |||
GenBank mRNA | AK033771.1 | - | 1 | 1400 | ||
AK080832.1 | - | 1 | 564 | |||
AK160135.1 | - | 1 | 491 | |||
BC002221.1 | - | 1 | 224 | |||
BC099379.1 | - | 1 | 539 |

More Information
Other Species Targeted

Gene Ontology Categories:
