siRNA Details
Gene Symbol: Celf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244891.1 | NP_001231820.1 | 15 | 7356 | ||
NM_001244903.1 | NP_001231832.1 | 14 | 7303 | |||
NM_017368.3 | NP_059064.2 | 15 | 4380 | |||
NM_198683.2 | NP_941955.1 | 14 | 4228 | |||
GenBank mRNA | AK080832.1 | - | 1 | 534 | ||
AK160135.1 | - | 1 | 461 | |||
BC002221.1 | - | 1 | 194 | |||
BC099379.1 | - | 1 | 509 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
