siRNA Details
Gene Symbol: Abr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291186.1 | NP_001278115.1 | 8 | 946 | ||
NM_198018.2 | NP_932135.1 | 12 | 1721 | |||
NM_198894.2 | NP_942597.1 | 13 | 1607 | |||
NM_198895.2 | NP_942598.1 | 13 | 1728 | |||
GenBank mRNA | AK134993.1 | - | 8 | 934 | ||
AK153615.1 | - | 12 | 2389 | |||
BC056385.1 | - | 8 | 935 | 6854368 | ||
BC058708.1 | - | 12 | 1721 | 6839300 | ||
BC059064.1 | - | 8 | 930 | 6825281 |
