siRNA Details
Gene Symbol: Serpinb6e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253875.1 | XP_006253937.1 | 3 | 460 |
Gene Symbol: RGD1564786 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253873.2 | XP_006253935.2 | 3 | 600 | ||
XM_008771602.1 | XP_008769824.1 | 4 | 799 | |||
XM_008771603.1 | XP_008769825.1 | 2 | 447 |
Gene Symbol: Serpinb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199085.2 | NP_954516.1 | 4 | 528 | ||
XM_006253869.1 | XP_006253931.1 | 4 | 526 | |||
XM_006253870.2 | XP_006253932.1 | 4 | 821 | |||
XM_006253871.2 | XP_006253933.1 | 4 | 641 | |||
XM_008771596.1 | XP_008769818.1 | 4 | 739 | |||
GenBank mRNA | BC060594.1 | - | 4 | 532 | 6887824 |
Gene Symbol: Serpinb6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012214.1 | NP_001012214.1 | 4 | 450 | ||
GenBank mRNA | BC079108.1 | - | 4 | 450 | 7125468 |

More Information
Other Species Targeted

Gene Ontology Categories:
