siRNA Details
Gene Symbol: ACTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101.3 | NP_001092.1 | 6 | 1470 | ||
GenBank mRNA | AK025375.1 | - | 6 | 1469 | ||
AK058019.1 | - | 6 | 1470 | |||
AK098751.1 | - | 7 | 1273 | |||
AK130157.1 | - | 6 | 1471 | |||
AK223032.1 | - | 6 | 1468 | |||
AK309997.1 | - | 7 | 1512 | |||
BC001301.1 | - | 6 | 1453 | 3451917 | ||
BC004251.1 | - | 6 | 1447 | 3623067 | ||
BC009636.1 | - | 1 | 204 | |||
BC012854.1 | - | 5 | 1341 | |||
BC013380.2 | - | 6 | 1462 | 4025526 | ||
BC014861.1 | - | 6 | 1460 | 3870487 | ||
X63432.1 | - | 6 | 1458 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
