siRNA Details
Gene Symbol: ACTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101.3 | NP_001092.1 | 6 | 1585 | ||
GenBank mRNA | AK025375.1 | - | 6 | 1584 | ||
AK058019.1 | - | 6 | 1585 | |||
AK098751.1 | - | 7 | 1388 | |||
AK130062.1 | - | 6 | 1587 | |||
AK130157.1 | - | 6 | 1586 | |||
AK223032.1 | - | 6 | 1583 | |||
AK223055.1 | - | 6 | 1583 | |||
AK225414.1 | - | 6 | 1420 | |||
AK309997.1 | - | 7 | 1627 | |||
BC001301.1 | - | 6 | 1568 | 3451917 | ||
BC002409.2 | - | 6 | 1573 | 2961617 | ||
BC004251.1 | - | 6 | 1562 | 3623067 | ||
BC008633.1 | - | 5 | 1482 | |||
BC009636.1 | - | 1 | 319 | |||
BC012854.1 | - | 5 | 1456 | |||
BC013380.2 | - | 6 | 1577 | 4025526 | ||
BC013835.1 | - | 6 | 1574 | |||
BC014861.1 | - | 6 | 1575 | 3870487 | ||
BC016045.1 | - | 6 | 1576 | 4564532 | ||
V00478.1 | - | 3 | 748 | |||
X00351.1 | - | 6 | 1542 | |||
X63432.1 | - | 6 | 1573 |
Gene Symbol: LOC644936 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004845.1 | 1 | 1166 | |||
GenBank mRNA | BC092424.1 | - | 1 | 1166 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
