siRNA Details
Gene Symbol: ACSM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308169.1 | NP_001295098.1 | 13 | 1789 | ||
NM_001308172.1 | NP_001295101.1 | 14 | 1974 | |||
NM_001308954.1 | NP_001295883.1 | 15 | 2356 | |||
XM_017022923.1 | XP_016878412.1 | 15 | 1949 | |||
XM_017022926.1 | XP_016878415.1 | 12 | 1491 | |||
GenBank mRNA | AK055867.1 | - | 15 | 1770 | ||
AK091878.1 | - | 7 | 1940 | |||
AK096039.1 | - | 15 | 2215 | |||
AK298359.1 | - | 13 | 1678 | |||
BC125176.1 | - | 14 | 1811 | 40121949 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
