siRNA Details
Gene Symbol: ACSM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308169.1 | NP_001295098.1 | 13 | 1936 | ||
NM_001308172.1 | NP_001295101.1 | 14 | 2121 | |||
NM_001308954.1 | NP_001295883.1 | 15 | 2503 | |||
XM_017022923.1 | XP_016878412.1 | 15 | 2096 | |||
XM_017022926.1 | XP_016878415.1 | 12 | 1638 | |||
GenBank mRNA | AK055867.1 | - | 15 | 1917 | ||
AK091878.1 | - | 7 | 2087 | |||
AK298359.1 | - | 13 | 1825 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
