siRNA Details
Gene Symbol: PRKAR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276289.1 | NP_001263218.1 | 3 | 380 | ||
NM_001276290.1 | NP_001263219.1 | 1 | 77 | |||
NM_001278433.1 | NP_001265362.1 | 2 | 250 | |||
NM_002734.4 | NP_002725.1 | 2 | 247 | |||
NM_212471.2 | NP_997636.1 | 2 | 440 | |||
NM_212472.2 | NP_997637.1 | 2 | 278 | |||
XM_011524983.2 | XP_011523285.1 | 2 | 343 | |||
XM_011524984.2 | XP_011523286.1 | 3 | 281 | |||
XM_011524985.2 | XP_011523287.1 | 3 | 575 | |||
GenBank mRNA | A12295.1 | - | 2 | 173 | ||
AA015682.1 | - | 2 | 214 | |||
AK097580.1 | - | 1 | 650 | |||
AK124586.1 | - | 2 | 176 | |||
AK312307.1 | - | 2 | 176 | |||
AL535889.3 | - | 2 | 218 | |||
BC036285.1 | - | 2 | 199 | 4340015 | ||
BC093042.1 | - | 2 | 188 | 6514677 | ||
BM928493.1 | - | 2 | 245 | |||
BU167718.1 | - | 2 | 287 | |||
BU188258.1 | - | 2 | 240 | |||
CR749311.1 | - | 3 | 309 | |||
DA108801.1 | - | 2 | 250 | |||
DA219203.1 | - | 2 | 247 | |||
M18468.1 | - | 2 | 174 | |||
M33336.1 | - | 2 | 158 | |||
S54705.1 | - | 2 | 185 | |||
Y07642.1 | - | 2 | 220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
