siRNA Details
Gene Symbol: PRKAR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276289.1 | NP_001263218.1 | 8 | 958 | ||
NM_001276290.1 | NP_001263219.1 | 6 | 655 | |||
NM_001278433.1 | NP_001265362.1 | 7 | 828 | |||
NM_002734.4 | NP_002725.1 | 7 | 825 | |||
NM_212471.2 | NP_997636.1 | 7 | 1018 | |||
NM_212472.2 | NP_997637.1 | 7 | 856 | |||
XM_011524983.2 | XP_011523285.1 | 7 | 921 | |||
XM_011524984.2 | XP_011523286.1 | 8 | 859 | |||
XM_011524985.2 | XP_011523287.1 | 8 | 1153 | |||
GenBank mRNA | A12295.1 | - | 7 | 751 | ||
AK097580.1 | - | 6 | 1228 | |||
AK124586.1 | - | 7 | 744 | |||
AK312307.1 | - | 7 | 754 | |||
AL535889.3 | - | 7 | 796 | |||
BC036285.1 | - | 7 | 777 | 4340015 | ||
BC093042.1 | - | 7 | 766 | 6514677 | ||
CR749311.1 | - | 8 | 887 | |||
M18468.1 | - | 7 | 752 | |||
M33336.1 | - | 7 | 736 | |||
S54707.1 | - | 3 | 181 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_021880, HQ412667, AK146380, BC005697, AK005039, and others.
Gene Ontology Categories:
