siRNA Details
Gene Symbol: DTX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300727.1 | NP_001287656.1 | 2 | 728 | ||
NM_015177.1 | NP_055992.1 | 2 | 1137 | |||
XM_006718483.3 | XP_006718546.1 | 2 | 1181 | |||
GenBank mRNA | AB023154.1 | - | 2 | 984 | ||
AF086290.1 | - | 1 | 283 | |||
BC119011.1 | - | 2 | 728 | 40085251 | ||
BC122861.1 | - | 2 | 728 | 40085252 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
