siRNA Details
Gene Symbol: DTX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300727.1 | NP_001287656.1 | 9 | 5166 | ||
NM_015177.1 | NP_055992.1 | 9 | 5575 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
