siRNA Details
Gene Symbol: CTCFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC137482.1 | - | 1 | 249 | 9021482 | |
BC137483.1 | - | 1 | 249 | 9021483 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
