siRNA Details
Gene Symbol: KIFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002263.3 | NP_002254.2 | 7 | 1836 | ||
XM_011514585.1 | XP_011512887.1 | 7 | 1542 | |||
XM_017010836.1 | XP_016866325.1 | 7 | 1542 | |||
XM_017010837.1 | XP_016866326.1 | 7 | 1651 | |||
GenBank mRNA | AK297378.1 | - | 7 | 1449 | ||
AK303239.1 | - | 7 | 1476 | |||
BC000712.2 | - | 7 | 1544 | |||
BC063567.1 | - | 7 | 1494 | |||
BC073878.1 | - | 7 | 1529 | |||
BC098438.1 | - | 7 | 1526 | |||
BC121041.1 | - | 7 | 1467 | 40117894 | ||
BC121042.1 | - | 7 | 1467 | 40117899 | ||
D14678.1 | - | 2 | 926 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
