siRNA Details
Gene Symbol: HIST1H4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003544.2 | NP_003535.1 | 1 | 27 | ||
GenBank mRNA | BC069467.1 | - | 1 | 27 | 7262716 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
