siRNA Details
Gene Symbol: HIST1H2BJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021058.3 | NP_066402.2 | 1 | 245 | ||
GenBank mRNA | AK311849.1 | - | 1 | 273 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
