siRNA Details
Gene Symbol: NT5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014595.2 | NP_055410.1 | 4 | 474 | ||
NR_045513.1 | 4 | 530 | ||||
XM_011524700.2 | XP_011523002.2 | 3 | 583 | |||
GenBank mRNA | AF154829.2 | - | 4 | 461 | ||
AK000419.1 | - | 4 | 461 | |||
BC008183.1 | - | 4 | 478 | 4183256 | ||
BC017454.1 | - | 4 | 439 | 4647090 | ||
BC022334.1 | - | 4 | 421 | 4703567 | ||
BQ431723.1 | - | 4 | 474 | |||
EU794640.1 | - | 4 | 461 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
